5-Bromo[1,1'-biphenyl]-2-yl-3-piperidinyl ether hydrochloride

Supplier: Matrix Scientific

050427-500MG 050427-2.5G
101891-608EA 364.62 USD
101891-608 101891-610
5-Bromo[1,1'-biphenyl]-2-yl-3-piperidinyl ether hydrochloride
5-Bromo[1,1'-biphenyl]-2-yl-3-piperidinyl ether hydrochloride

Hazard: Irritant

Formula: C₁₇H₁₉BrClNO
MDL Number: MFCD13560828
CAS Number: 1220030-61-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR