3-[2-Bromo-4-(1-methyl-1-phenylethyl)phenoxy]pyrrolidine hydrochloride
Supplier: Matrix Scientific
050426-500MG
050426-2.5G
101891-604EA
364.62
USD
101891-604
101891-606
3-[2-Bromo-4-(1-methyl-1-phenylethyl)phenoxy]pyrrolidine hydrochloride
3-[2-Bromo-4-(1-methyl-1-phenylethyl)phenoxy]pyrrolidine hydrochloride
Hazard: Irritant
Formula:
C₁₉H₂₃BrClNO |
MDL Number:
MFCD13560827 CAS Number: 1220030-66-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...