4-[2-Bromo-4-(1-methyl-1-phenylethyl)phenoxy]piperidine hydrochloride

Supplier: Matrix Scientific

050423-500MG 050423-2.5G
101891-592EA 364.62 USD
101891-592 101891-594
4-[2-Bromo-4-(1-methyl-1-phenylethyl)phenoxy]piperidine hydrochloride
4-[2-Bromo-4-(1-methyl-1-phenylethyl)phenoxy]piperidine hydrochloride

Hazard: Irritant

Formula: C₂₀H₂₅BrClNO
MDL Number: MFCD13560824
CAS Number: 1220027-12-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR