3-(4-Chloro-2-methylphenoxy)pyrrolidine hydrochloride

Supplier: Matrix Scientific

050421-500MG 050421-2.5G
101891-584EA 364.62 USD
101891-584 101891-586
3-(4-Chloro-2-methylphenoxy)pyrrolidine hydrochloride
3-(4-Chloro-2-methylphenoxy)pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₅Cl₂NO
MDL Number: MFCD13560822
CAS Number: 1219971-75-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR