4-(4-Chloro-2-methylphenoxy)piperidine hydrochloride

Supplier: Matrix Scientific

050418-500MG 050418-2.5G
101891-572EA 364.62 USD
101891-572 101891-574
4-(4-Chloro-2-methylphenoxy)piperidine hydrochloride
4-(4-Chloro-2-methylphenoxy)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇Cl₂NO
MDL Number: MFCD13560819
CAS Number: 921204-98-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR