3-(4-Chloro-2-methylphenoxy)piperidine hydrochloride

Supplier: Matrix Scientific

050417-500MG 050417-2.5G
101891-568EA 364.62 USD
101891-568 101891-570
3-(4-Chloro-2-methylphenoxy)piperidine hydrochloride
3-(4-Chloro-2-methylphenoxy)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇Cl₂NO
MDL Number: MFCD13560818
CAS Number: 1219968-07-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR