3-(4-Bromo-3-methylphenoxy)pyrrolidine hydrochloride

Supplier: Matrix Scientific

050416-500MG 050416-2.5G
101891-564EA 364.62 USD
101891-564 101891-566
3-(4-Bromo-3-methylphenoxy)pyrrolidine hydrochloride
3-(4-Bromo-3-methylphenoxy)pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₅BrClNO
MDL Number: MFCD13560817
CAS Number: 1220027-23-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR