4-[(4-Bromo-3-methylphenoxy)methyl]piperidine hydrochloride

Supplier: Matrix Scientific

050415-500MG 050415-2.5G
101891-560EA 364.62 USD
101891-560 101891-562
4-[(4-Bromo-3-methylphenoxy)methyl]piperidine hydrochloride
4-[(4-Bromo-3-methylphenoxy)methyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉BrClNO
MDL Number: MFCD13560816
CAS Number: 1220032-80-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR