3-[(4-Bromo-3-methylphenoxy)methyl]piperidine hydrochloride

Supplier: Matrix Scientific

050414-500MG 050414-2.5G
101891-556EA 364.62 USD
101891-556 101891-558
3-[(4-Bromo-3-methylphenoxy)methyl]piperidine hydrochloride
3-[(4-Bromo-3-methylphenoxy)methyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉BrClNO
MDL Number: MFCD13560815
CAS Number: 1220027-03-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR