3-(2-Bromo-4-ethylphenoxy)pyrrolidine hydrochloride
Supplier: Matrix Scientific
050411-500MG
050411-2.5G
101891-544EA
364.62
USD
101891-544
101891-546
3-(2-Bromo-4-ethylphenoxy)pyrrolidine hydrochloride
3-(2-Bromo-4-ethylphenoxy)pyrrolidine hydrochloride
Hazard: Irritant
Formula:
C₁₂H₁₇BrClNO |
MDL Number:
MFCD13560812 CAS Number: 1219967-92-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...