4-(2-Bromo-4-ethylphenoxy)piperidine hydrochloride

Supplier: Matrix Scientific

050408-500MG 050408-2.5G
101891-532EA 364.62 USD
101891-532 101891-534
4-(2-Bromo-4-ethylphenoxy)piperidine hydrochloride
4-(2-Bromo-4-ethylphenoxy)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉BrClNO
MDL Number: MFCD13560809
CAS Number: 1220032-78-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR