3-(2-Bromo-4-ethylphenoxy)piperidine hydrochloride

Supplier: Matrix Scientific

050407-500MG 050407-2.5G
101891-528EA 364.62 USD
101891-528 101891-530
3-(2-Bromo-4-ethylphenoxy)piperidine hydrochloride
3-(2-Bromo-4-ethylphenoxy)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉BrClNO
MDL Number: MFCD13560808
CAS Number: 1219967-88-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR