3-[2-Bromo-4-(tert-butyl)phenoxy]pyrrolidine hydrochloride

Supplier: Matrix Scientific

050406-500MG 050406-2.5G
101891-524EA 364.62 USD
101891-524 101891-526
3-[2-Bromo-4-(tert-butyl)phenoxy]pyrrolidine hydrochloride
3-[2-Bromo-4-(tert-butyl)phenoxy]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₁BrClNO
MDL Number: MFCD13560807
CAS Number: 1219967-74-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR