2-Bromo-4-(tert-butyl)phenyl-4-piperidinylmethylether hydrochloride

Supplier: Matrix Scientific

050405-500MG 050405-2.5G
101891-520EA 364.62 USD
101891-520 101891-522
2-Bromo-4-(tert-butyl)phenyl-4-piperidinylmethylether hydrochloride
2-Bromo-4-(tert-butyl)phenyl-4-piperidinylmethylether hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₅BrClNO
MW: 362.75 g/mol
MDL Number: MFCD13560806
CAS Number: 1219967-81-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR