2-Bromo-4-(tert-butyl)phenyl-3-piperidinylmethylether hydrochloride

Supplier: Matrix Scientific

050404-500MG 050404-2.5G
101891-516EA 364.62 USD
101891-516 101891-518
2-Bromo-4-(tert-butyl)phenyl-3-piperidinylmethylether hydrochloride
2-Bromo-4-(tert-butyl)phenyl-3-piperidinylmethylether hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₅BrClNO
MW: 362.75 g/mol
MDL Number: MFCD13560805
CAS Number: 1220030-42-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR