4-[2-Bromo-4-(tert-butyl)phenoxy]piperidine hydrochloride

Supplier: Matrix Scientific

050403-500MG 050403-2.5G
101891-512EA 364.62 USD
101891-512 101891-514
4-[2-Bromo-4-(tert-butyl)phenoxy]piperidine hydrochloride
4-[2-Bromo-4-(tert-butyl)phenoxy]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₃BrClNO
MDL Number: MFCD13560804
CAS Number: 1220021-49-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR