3-[2-Bromo-4-(tert-butyl)phenoxy]piperidine hydrochloride

Supplier: Matrix Scientific

050402-500MG 050402-2.5G
101891-508EA 364.62 USD
101891-508 101891-510
3-[2-Bromo-4-(tert-butyl)phenoxy]piperidine hydrochloride
3-[2-Bromo-4-(tert-butyl)phenoxy]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₃BrClNO
MDL Number: MFCD13560803
CAS Number: 1220019-29-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR