4-{[(3-Bromo[1,1'-biphenyl]-4-yl)oxy]methyl}piperidine hydrochloride

Supplier: Matrix Scientific

050400-500MG 050400-2.5G
101891-500EA 364.62 USD
101891-500 101891-502
4-{[(3-Bromo[1,1'-biphenyl]-4-yl)oxy]methyl}piperidine hydrochloride
4-{[(3-Bromo[1,1'-biphenyl]-4-yl)oxy]methyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₈H₂₁BrClNO
MDL Number: MFCD13560801
CAS Number: 1220018-05-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR