3-Bromo[1,1'-biphenyl]-4-yl-4-piperidinyl ether hydrochloride

Supplier: Matrix Scientific

050398-500MG 050398-2.5G
101891-492EA 186.15 USD
101891-492 101891-494
3-Bromo[1,1'-biphenyl]-4-yl-4-piperidinyl ether hydrochloride
3-Bromo[1,1'-biphenyl]-4-yl-4-piperidinyl ether hydrochloride

Hazard: Irritant

Formula: C₁₇H₁₉BrClNO
MDL Number: MFCD13560799
CAS Number: 1220035-27-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR