3-Bromo[1,1'-biphenyl]-4-yl-3-piperidinyl ether hydrochloride

Supplier: Matrix Scientific

050397-500MG 050397-2.5G
101891-488EA 364.62 USD
101891-488 101891-490
3-Bromo[1,1'-biphenyl]-4-yl-3-piperidinyl ether hydrochloride
3-Bromo[1,1'-biphenyl]-4-yl-3-piperidinyl ether hydrochloride

Hazard: Irritant

Formula: C₁₇H₁₉BrClNO
MDL Number: MFCD13560798
CAS Number: 1220021-40-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR