3-(2-Bromo-4-isopropylphenoxy)pyrrolidine hydrochloride

Supplier: Matrix Scientific

050396-500MG 050396-2.5G
101891-484EA 364.62 USD
101891-484 101891-486
3-(2-Bromo-4-isopropylphenoxy)pyrrolidine hydrochloride
3-(2-Bromo-4-isopropylphenoxy)pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉BrClNO
MDL Number: MFCD13560797
CAS Number: 1220032-76-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR