3-[(2-Bromo-4-isopropylphenoxy)methyl]piperidine hydrochloride

Supplier: Matrix Scientific

050394-500MG 050394-2.5G
101891-476EA 364.62 USD
101891-476 101891-478
3-[(2-Bromo-4-isopropylphenoxy)methyl]piperidine hydrochloride
3-[(2-Bromo-4-isopropylphenoxy)methyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₃BrClNO
MDL Number: MFCD13560795
CAS Number: 1219972-63-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR