4-(2-Bromo-4-isopropylphenoxy)piperidine hydrochloride

Supplier: Matrix Scientific

050393-500MG 050393-2.5G
101891-472EA 364.62 USD
101891-472 101891-474
4-(2-Bromo-4-isopropylphenoxy)piperidine hydrochloride
4-(2-Bromo-4-isopropylphenoxy)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₁BrClNO
MDL Number: MFCD13560794
CAS Number: 1220019-07-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR