3-(2-Bromo-4-isopropylphenoxy)piperidine hydrochloride
Supplier: Matrix Scientific
050392-500MG
050392-2.5G
101891-468EA
364.62
USD
101891-468
101891-470
3-(2-Bromo-4-isopropylphenoxy)piperidine hydrochloride
3-(2-Bromo-4-isopropylphenoxy)piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₄H₂₁BrClNO |
MDL Number:
MFCD13560793 CAS Number: 1220019-15-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...