4-{[2,4-Di(tert-butyl)phenoxy]methyl}piperidine hydrochloride

Supplier: Matrix Scientific

050390-500MG 050390-2.5G
101891-460EA 364.62 USD
101891-460 101891-462
4-{[2,4-Di(tert-butyl)phenoxy]methyl}piperidine hydrochloride
4-{[2,4-Di(tert-butyl)phenoxy]methyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₂₀H₃₄ClNO
MDL Number: MFCD13560791
CAS Number: 1220021-32-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR