3-{[2,4-Di(tert-butyl)phenoxy]methyl}piperidine hydrochloride

Supplier: Matrix Scientific

050389-500MG 050389-2.5G
101891-456EA 364.62 USD
101891-456 101891-458
3-{[2,4-Di(tert-butyl)phenoxy]methyl}piperidine hydrochloride
3-{[2,4-Di(tert-butyl)phenoxy]methyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₂₀H₃₄ClNO
MDL Number: MFCD13560790
CAS Number: 1219972-58-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR