4-[2,4-Di(tert-butyl)phenoxy]piperidine hydrochloride
Supplier: Matrix Scientific
050388-500MG
050388-2.5G
101891-452EA
364.62
USD
101891-452
101891-454
4-[2,4-Di(tert-butyl)phenoxy]piperidine hydrochloride
4-[2,4-Di(tert-butyl)phenoxy]piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₉H₃₂ClNO |
MDL Number:
MFCD13560789 CAS Number: 1220021-20-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...