3-[2,4-Di(tert-butyl)phenoxy]piperidine hydrochloride

Supplier: Matrix Scientific

050387-500MG 050387-2.5G
101891-448EA 364.62 USD
101891-448 101891-450
3-[2,4-Di(tert-butyl)phenoxy]piperidine hydrochloride
3-[2,4-Di(tert-butyl)phenoxy]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₉H₃₂ClNO
MDL Number: MFCD13560788
CAS Number: 1220032-75-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR