3-[4-Bromo-2-(tert-butyl)phenoxy]pyrrolidine hydrochloride

Supplier: Matrix Scientific

050386-500MG 050386-2.5G
101891-444EA 364.62 USD
101891-444 101891-446
3-[4-Bromo-2-(tert-butyl)phenoxy]pyrrolidine hydrochloride
3-[4-Bromo-2-(tert-butyl)phenoxy]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₁BrClNO
MDL Number: MFCD13560787
CAS Number: 1220017-73-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR