4-Bromo-2-(tert-butyl)phenyl-4-piperidinylmethylether hydrochloride

Supplier: Matrix Scientific

050385-500MG 050385-2.5G
101891-440EA 364.62 USD
101891-440 101891-442
4-Bromo-2-(tert-butyl)phenyl-4-piperidinylmethylether hydrochloride
4-Bromo-2-(tert-butyl)phenyl-4-piperidinylmethylether hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₅BrClNO
MW: 362.75 g/mol
MDL Number: MFCD13560786
CAS Number: 1220018-95-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR