4-[4-Bromo-2-(tert-butyl)phenoxy]piperidine hydrochloride

Supplier: Matrix Scientific

050383-500MG 050383-2.5G
101891-432EA 364.62 USD
101891-432 101891-434
4-[4-Bromo-2-(tert-butyl)phenoxy]piperidine hydrochloride
4-[4-Bromo-2-(tert-butyl)phenoxy]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₃BrClNO
MDL Number: MFCD13560784
CAS Number: 1220021-09-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR