3-[4-Bromo-2-(tert-butyl)phenoxy]piperidine hydrochloride

Supplier: Matrix Scientific

050382-500MG 050382-2.5G
101891-428EA 364.62 USD
101891-428 101891-430
3-[4-Bromo-2-(tert-butyl)phenoxy]piperidine hydrochloride
3-[4-Bromo-2-(tert-butyl)phenoxy]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₃BrClNO
MDL Number: MFCD13560783
CAS Number: 1220018-83-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR