3-(4-Bromo-2-isopropylphenoxy)pyrrolidine hydrochloride

Supplier: Matrix Scientific

050381-500MG 050381-2.5G
101891-424EA 364.62 USD
101891-424 101891-426
3-(4-Bromo-2-isopropylphenoxy)pyrrolidine hydrochloride
3-(4-Bromo-2-isopropylphenoxy)pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉BrClNO
MDL Number: MFCD13560782
CAS Number: 1220032-74-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR