4-[(4-Bromo-2-isopropylphenoxy)methyl]piperidine hydrochloride

Supplier: Matrix Scientific

050380-500MG 050380-2.5G
101891-420EA 364.62 USD
101891-420 101891-422
4-[(4-Bromo-2-isopropylphenoxy)methyl]piperidine hydrochloride
4-[(4-Bromo-2-isopropylphenoxy)methyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₃BrClNO
MDL Number: MFCD13560781
CAS Number: 1219972-50-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR