4-(4-Bromo-2-isopropylphenoxy)piperidine hydrochloride
Supplier: Matrix Scientific
050378-500MG
050378-2.5G
101891-412EA
364.62
USD
101891-412
101891-414
4-(4-Bromo-2-isopropylphenoxy)piperidine hydrochloride
4-(4-Bromo-2-isopropylphenoxy)piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₄H₂₁BrClNO |
MDL Number:
MFCD13560779 CAS Number: 1220032-72-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...