3-[2-Bromo-4-(sec-butyl)phenoxy]pyrrolidine hydrochloride

Supplier: Matrix Scientific

050376-500MG 050376-2.5G
101891-404EA 364.62 USD
101891-404 101891-406
3-[2-Bromo-4-(sec-butyl)phenoxy]pyrrolidine hydrochloride
3-[2-Bromo-4-(sec-butyl)phenoxy]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₁BrClNO
MDL Number: MFCD13560777
CAS Number: 1219972-45-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR