4-{[2-Bromo-4-(sec-butyl)phenoxy]methyl}piperidine hydrochloride

Supplier: Matrix Scientific

050375-500MG 050375-2.5G
101891-400EA 364.62 USD
101891-400 101891-402
4-{[2-Bromo-4-(sec-butyl)phenoxy]methyl}piperidine hydrochloride
4-{[2-Bromo-4-(sec-butyl)phenoxy]methyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₅BrClNO
MDL Number: MFCD13560776
CAS Number: 1220020-92-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR