3-{[2-Bromo-4-(sec-butyl)phenoxy]methyl}piperidine hydrochloride

Supplier: Matrix Scientific

050374-500MG 050374-2.5G
101891-396EA 364.62 USD
101891-396 101891-398
3-{[2-Bromo-4-(sec-butyl)phenoxy]methyl}piperidine hydrochloride
3-{[2-Bromo-4-(sec-butyl)phenoxy]methyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₅BrClNO
MDL Number: MFCD13560775
CAS Number: 1220018-61-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR