4-[2-Bromo-4-(sec-butyl)phenoxy]piperidine hydrochloride

Supplier: Matrix Scientific

050373-500MG 050373-2.5G
101891-392EA 364.62 USD
101891-392 101891-394
4-[2-Bromo-4-(sec-butyl)phenoxy]piperidine hydrochloride
4-[2-Bromo-4-(sec-butyl)phenoxy]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₃BrClNO
MDL Number: MFCD13560774
CAS Number: 1220018-72-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR