3-[2-Bromo-4-(sec-butyl)phenoxy]piperidine hydrochloride

Supplier: Matrix Scientific

050372-500MG 050372-2.5G
101891-388EA 364.62 USD
101891-388 101891-390
3-[2-Bromo-4-(sec-butyl)phenoxy]piperidine hydrochloride
3-[2-Bromo-4-(sec-butyl)phenoxy]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₃BrClNO
MDL Number: MFCD13560773
CAS Number: 1220032-73-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR