3-[(1,6-Dibromo-2-naphthyl)oxy]pyrrolidine hydrochloride

Supplier: Matrix Scientific

050371-500MG 050371-2.5G
101891-384EA 364.62 USD
101891-384 101891-386
3-[(1,6-Dibromo-2-naphthyl)oxy]pyrrolidine hydrochloride
3-[(1,6-Dibromo-2-naphthyl)oxy]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₁₄Br₂ClNO
MDL Number: MFCD13560772
CAS Number: 1219972-43-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR