4-{[(1,6-Dibromo-2-naphthyl)oxy]methyl}piperidine hydrochloride

Supplier: Matrix Scientific

050370-500MG 050370-2.5G
101891-380EA 364.62 USD
101891-380 101891-382
4-{[(1,6-Dibromo-2-naphthyl)oxy]methyl}piperidine hydrochloride
4-{[(1,6-Dibromo-2-naphthyl)oxy]methyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₆H₁₈Br₂ClNO
MDL Number: MFCD13560771
CAS Number: 1220035-26-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR