4-[(1,6-Dibromo-2-naphthyl)oxy]piperidine hydrochloride

Supplier: Matrix Scientific

050368-500MG 050368-2.5G
101891-372EA 364.62 USD
101891-372 101891-374
4-[(1,6-Dibromo-2-naphthyl)oxy]piperidine hydrochloride
4-[(1,6-Dibromo-2-naphthyl)oxy]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₁₆Br₂ClNO
MDL Number: MFCD13560769
CAS Number: 1220018-23-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR