3-[(2,4-Dichloro-1-naphthyl)oxy]pyrrolidine hydrochloride

Supplier: Matrix Scientific

050366-500MG 050366-2.5G
101891-364EA 364.62 USD
101891-364 101891-366
3-[(2,4-Dichloro-1-naphthyl)oxy]pyrrolidine hydrochloride
3-[(2,4-Dichloro-1-naphthyl)oxy]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₁₄Cl₃NO
MDL Number: MFCD13560767
CAS Number: 1219972-38-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR