2,4-Dichloro-1-naphthyl-4-piperidinylmethyl ether hydrochloride
Supplier: Matrix Scientific
050365-500MG
050365-2.5G
101891-360EA
364.62
USD
101891-360
101891-362
2,4-Dichloro-1-naphthyl-4-piperidinylmethyl ether hydrochloride
2,4-Dichloro-1-naphthyl-4-piperidinylmethyl ether hydrochloride
Hazard: Irritant
Formula:
C₁₆H₁₈Cl₃NO MW: 346.69 g/mol |
MDL Number:
MFCD13560766 CAS Number: 1220032-69-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...