2,4-Dichloro-1-naphthyl-3-piperidinylmethyl ether hydrochloride

Supplier: Matrix Scientific

050364-500MG 050364-2.5G
101891-356EA 364.62 USD
101891-356 101891-358
2,4-Dichloro-1-naphthyl-3-piperidinylmethyl ether hydrochloride
2,4-Dichloro-1-naphthyl-3-piperidinylmethyl ether hydrochloride

Hazard: Irritant

Formula: C₁₆H₁₈Cl₃NO
MDL Number: MFCD13560765
CAS Number: 1219972-40-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR