3-(4-Bromo-2-propylphenoxy)pyrrolidine hydrochloride

Supplier: Matrix Scientific

050361-500MG 050361-2.5G
101891-344EA 364.62 USD
101891-344 101891-346
3-(4-Bromo-2-propylphenoxy)pyrrolidine hydrochloride
3-(4-Bromo-2-propylphenoxy)pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉BrClNO
MDL Number: MFCD13560762
CAS Number: 1220018-08-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR