3-[(4-Bromo-2-propylphenoxy)methyl]piperidine hydrochloride

Supplier: Matrix Scientific

050359-500MG 050359-2.5G
101891-336EA 364.62 USD
101891-336 101891-338
3-[(4-Bromo-2-propylphenoxy)methyl]piperidine hydrochloride
3-[(4-Bromo-2-propylphenoxy)methyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₃BrClNO
MDL Number: MFCD13560760
CAS Number: 1220032-67-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR