4-(4-Bromo-2-propylphenoxy)piperidine hydrochloride

Supplier: Matrix Scientific

050358-500MG 050358-2.5G
101891-332EA 364.62 USD
101891-332 101891-334
4-(4-Bromo-2-propylphenoxy)piperidine hydrochloride
4-(4-Bromo-2-propylphenoxy)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₁BrClNO
MDL Number: MFCD13560759
CAS Number: 1220020-86-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR