3-(2-Bromo-4-chloro-3,5-dimethylphenoxy)pyrrolidine hydrochloride

Supplier: Matrix Scientific

050356-500MG 050356-2.5G
101891-324EA 364.62 USD
101891-324 101891-326
3-(2-Bromo-4-chloro-3,5-dimethylphenoxy)pyrrolidine hydrochloride
3-(2-Bromo-4-chloro-3,5-dimethylphenoxy)pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₆BrCl₂NO
MDL Number: MFCD13560757
CAS Number: 1220018-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR